siRNA Details
| Gene Symbol: Zfp935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136496.1 | NP_001129968.1 | 3 | 1496 | ||
| NM_178875.4 | NP_849206.2 | 4 | 1557 | |||
| GenBank mRNA | AK018441.1 | - | 3 | 1487 | ||
| AY189975.1 | - | 3 | 1473 | |||
| BC012524.1 | - | 3 | 1472 | 4008347 | ||
| BC037448.1 | - | 3 | 1501 | |||
| BC047396.1 | - | 4 | 1534 | 3979342 | ||
| BC087936.1 | - | 4 | 1557 | |||
| BC113193.1 | - | 4 | 1553 | 40062446 | ||
| BC115966.1 | - | 1415 | ||||

 Zfp935
Zfp935 Genomic Map
Genomic Map