siRNA Details
| Gene Symbol: Alg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243161.1 | NP_001230090.1 | 3 | 512 | ||
| NM_183142.4 | NP_898965.1 | 3 | 386 | |||
| GenBank mRNA | AB288237.1 | - | 3 | 506 | ||
| AB288238.1 | - | 3 | 510 | |||
| AK046475.1 | - | 3 | 388 | |||
| AK134312.1 | - | 3 | 487 | |||
| AK157746.1 | - | 3 | 512 | |||
| BC061469.1 | - | 3 | 495 | 6821552 | ||

Alg11
Genomic Map