siRNA Details
| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197944.1 | NP_922935.1 | 4 | 370 | ||
| GenBank mRNA | AK157000.1 | - | 403 | |||
| AY319653.1 | - | 4 | 370 | |||
| BC094291.1 | - | 3 | 346 | 5316178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsh2d| Gene Symbol: Hsh2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197944.1 | NP_922935.1 | 4 | 370 | ||
| GenBank mRNA | AK157000.1 | - | 403 | |||
| AY319653.1 | - | 4 | 370 | |||
| BC094291.1 | - | 3 | 346 | 5316178 | ||