siRNA Details
| Gene Symbol: RTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005619.4 | NP_005610.1 | 9, 10 | 1735 | ||
| NM_206900.2 | NP_996783.1 | 8, 9 | 1516 | |||
| NM_206901.2 | NP_996784.1 | 5, 6 | 558 | |||
| GenBank mRNA | AF004222.1 | - | 9, 10 | 1634 | ||
| AF004223.1 | - | 6, 7 | 784 | |||
| AF004224.1 | - | 5, 6 | 526 | |||
| AF038540.1 | - | 6, 7 | 965 | |||
| AK291967.1 | - | 8, 9 | 1497 | |||
| AK314158.1 | - | 8, 9 | 1499 | |||
| BC073874.1 | - | 9, 10 | 1677 | 5242827 | ||

RTN2
Genomic Map