siRNA Details
Gene Symbol: Ostn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198112.2 | NP_932780.1 | 5 | 880 | ||
GenBank mRNA | AY395730.1 | - | 5 | 880 | ||
AY573932.1 | - | 5 | 1032 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ostn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198112.2 | NP_932780.1 | 5 | 880 | ||
GenBank mRNA | AY395730.1 | - | 5 | 880 | ||
AY573932.1 | - | 5 | 1032 |