siRNA Details
| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146010.1 | NP_001139482.1 | 6, 7 | 769 | ||
| NM_199012.2 | NP_950177.2 | 6, 7 | 769 | |||
| GenBank mRNA | AK122368.1 | - | 6, 7 | 661 | ||
| AK140322.1 | - | 6, 7 | 748 | |||
| AK141949.1 | - | 6, 7 | 769 | |||
| AK156841.1 | - | 5, 6 | 595 | |||
| AK157054.1 | - | 6, 7 | 742 | |||
| BC016532.1 | - | 6, 7 | 768 | |||
| BC060647.1 | - | 6, 7 | 731 | 6854041 | ||

Fchsd2
Genomic Map