siRNA Details
| Gene Symbol: 3110062M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135611.1 | NP_001129083.1 | 4 | 2783 | ||
| NM_199145.2 | NP_954596.1 | 4 | 3108 | |||
| GenBank mRNA | AK042380.1 | - | 4 | 3064 | ||
| AK077501.1 | - | 538 | ||||
| AK083365.1 | - | 4 | 2753 | |||

3110062M04Rik
Genomic Map