siRNA Details
| Gene Symbol: Tmem106c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252153.1 | NP_001239082.1 | 5 | 634 | ||
| NM_201359.2 | NP_958747.1 | 5 | 802 | |||
| GenBank mRNA | AK134684.1 | - | 5 | 636 | ||
| AK181938.1 | - | 2 | 153 | |||
| AK186053.1 | - | 2 | 153 | |||
| AK187749.1 | - | 1 | 72 | |||
| BC046621.1 | - | 5 | 752 | 6308771 | ||

Tmem106c
Genomic Map