siRNA Details
| Gene Symbol: Jtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206924.2 | NP_996807.1 | 4 | 626 | ||
| GenBank mRNA | AB016490.1 | - | 4 | 571 | ||
| AK004469.1 | - | 4 | 538 | |||
| AK089048.1 | - | 4 | 538 | |||
| AK146337.1 | - | 4 | 627 | |||
| AK215601.1 | - | 2 | 140 | |||
| BC008139.1 | - | 4 | 590 | 3582930 | ||
| BG077727.2 | - | 3 | 192 | |||

Jtb
Genomic Map