siRNA Details
| Gene Symbol: Mdga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193266.1 | NP_001180195.2 | 2 | 1678 | ||
| NM_207010.2 | NP_996893.3 | 2 | 1678 | |||
| GenBank mRNA | AK137667.1 | - | 2 | 1679 | ||
| AY371925.1 | - | 2 | 332 | |||
| BC137901.1 | - | 2 | 331 | 8860920 | ||
| BC137902.1 | - | 2 | 331 | 8860921 | ||
| BC145021.1 | - | 2 | 331 | |||
| BC171938.1 | - | 2 | 331 | 9054224 | ||

Mdga2
Genomic Map