siRNA Details
| Gene Symbol: Tas2r115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207020.1 | NP_996903.1 | 1 | 917 | ||
| GenBank mRNA | BC138393.1 | - | 1 | 966 | 8861413 | |
| BC138398.1 | - | 1 | 966 | 8861418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r115| Gene Symbol: Tas2r115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207020.1 | NP_996903.1 | 1 | 917 | ||
| GenBank mRNA | BC138393.1 | - | 1 | 966 | 8861413 | |
| BC138398.1 | - | 1 | 966 | 8861418 | ||