siRNA Details
| Gene Symbol: SSX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278691.1 | NP_001265620.1 | 7 | 657 | ||
| NM_005635.3 | NP_005626.1 | 6 | 571 | |||
| GenBank mRNA | BC001003.2 | - | 6 | 516 | 3445470 | |
| BC125151.1 | - | 6 | 514 | 40120554 | ||
| BC128611.1 | - | 6 | 513 | 6159943 | ||
| BC133693.1 | - | 6 | 514 | 40120550 | ||
| BC150487.2 | - | 6 | 518 | 40120555 | ||
| BE777476.1 | - | 7 | 594 | |||
| BF211314.1 | - | 6 | 535 | |||
| X86174.1 | - | 6 | 526 | |||

SSX1
Genomic Map