siRNA Details
Gene Symbol: Prr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207203.2 | NP_997086.2 | 6 | 2271 | ||
GenBank mRNA | BC068157.1 | - | 7 | 2178 | 30363411 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207203.2 | NP_997086.2 | 6 | 2271 | ||
GenBank mRNA | BC068157.1 | - | 7 | 2178 | 30363411 |