siRNA Details
| Gene Symbol: Dyrk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207210.2 | NP_997093.2 | 15 | 2017 | ||
| GenBank mRNA | AK077117.1 | - | 15 | 2018 | ||
| BC052324.1 | - | 14 | 2062 | 6315342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dyrk4| Gene Symbol: Dyrk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207210.2 | NP_997093.2 | 15 | 2017 | ||
| GenBank mRNA | AK077117.1 | - | 15 | 2018 | ||
| BC052324.1 | - | 14 | 2062 | 6315342 | ||