siRNA Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206335.1 | NP_001193264.1 | 3 | 465 | ||
| NM_207217.4 | NP_997100.1 | 2 | 353 | |||
| GenBank mRNA | AK029427.1 | - | 2 | 253 | ||
| AK158049.1 | - | 447 | ||||
| AK170646.1 | - | 3 | 384 | |||
| BC046457.1 | - | 3 | 312 | 4222961 | ||

Itfg3
Genomic Map