siRNA Details
| Gene Symbol: Oard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289490.1 | NP_001276419.1 | 3 | 527 | ||
| NM_001289491.1 | NP_001276420.1 | 3 | 252 | |||
| NM_207219.4 | NP_997102.2 | 3 | 392 | |||
| GenBank mRNA | AK148136.1 | - | 3 | 514 | ||
| AK153512.1 | - | 3 | 315 | |||
| BC022574.1 | - | 2 | 171 | 3989373 | ||
| BC157987.1 | - | 3 | 277 | 9007303 | ||
| BY741306.1 | - | 3 | 514 | |||

Oard1
Genomic Map