siRNA Details
| Gene Symbol: Oard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289490.1 | NP_001276419.1 | 6 | 833 | ||
| NM_001289491.1 | NP_001276420.1 | 6 | 558 | |||
| NM_207219.4 | NP_997102.2 | 6 | 698 | |||
| GenBank mRNA | AK148136.1 | - | 6 | 820 | ||
| AK153512.1 | - | 6 | 621 | |||
| BC022574.1 | - | 5 | 477 | 3989373 | ||
| BC157987.1 | - | 6 | 583 | 9007303 | ||

Oard1
Genomic Map