siRNA Details
| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177360.1 | NP_001170831.1 | 3, 4 | 1000 | ||
| NM_001177361.1 | NP_001170832.1 | 5, 6 | 1173 | |||
| NM_001177362.1 | NP_001170833.1 | 5, 6 | 864 | |||
| NM_207220.2 | NP_997103.1 | 5, 6 | 1276 | |||
| GenBank mRNA | AK039612.1 | - | 5, 6 | 776 | ||
| AK142928.1 | - | 3, 4 | 598 | |||
| AK143450.1 | - | 5, 6 | 1174 | |||
| AK170308.1 | - | 5, 6 | 866 | |||
| BC048078.1 | - | 6, 7 | 1148 | 6308873 | ||

Gpr137
Genomic Map