siRNA Details
Gene Symbol: Lmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207222.2 | NP_997105.1 | 5 | 1088 | ||
GenBank mRNA | AK034177.1 | - | 5 | 1089 | ||
BC034128.1 | - | 3 | 786 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207222.2 | NP_997105.1 | 5 | 1088 | ||
GenBank mRNA | AK034177.1 | - | 5 | 1089 | ||
BC034128.1 | - | 3 | 786 |