siRNA Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290720.1 | NP_001277649.1 | 6 | 920 | ||
| NM_001290721.1 | NP_001277650.1 | 5 | 727 | |||
| NM_207228.3 | NP_997111.1 | 6 | 787 | |||
| GenBank mRNA | AF530050.3 | - | 5 | 732 | ||
| AK030254.1 | - | 6 | 920 | |||
| AK149243.1 | - | 2 | 2684 | |||
| AK163138.1 | - | 6 | 734 | |||
| AK170053.1 | - | 6 | 810 | |||
| AY618264.1 | - | 5 | 732 | |||
| BC053407.1 | - | 5 | 710 | |||
| BC066782.1 | - | 6 | 784 | 6705127 | ||

Tsga10
Genomic Map