siRNA Details
| Gene Symbol: Tsga10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290720.1 | NP_001277649.1 | 11 | 1469 | ||
| NM_001290721.1 | NP_001277650.1 | 10 | 1276 | |||
| NM_207228.3 | NP_997111.1 | 11 | 1336 | |||
| GenBank mRNA | AF530050.3 | - | 10 | 1281 | ||
| AK030254.1 | - | 11 | 1469 | |||
| AY618264.1 | - | 10 | 1281 | |||
| BC066782.1 | - | 11 | 1333 | 6705127 | ||

Tsga10
Genomic Map