siRNA Details
| Gene Symbol: Rasgrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166493.1 | NP_001159965.1 | 3 | 495 | ||
| NM_207246.4 | NP_997129.1 | 4 | 487 | |||
| GenBank mRNA | AB154824.1 | - | 2 | 182 | ||
| AK173041.1 | - | 3 | 380 | |||
| BC066069.1 | - | 2 | 125 | 6851971 | ||
| BC066127.1 | - | 4 | 412 | 30280332 | ||
| CD348455.1 | - | 2 | 135 | |||
| CN695386.1 | - | 3 | 477 | |||

Rasgrp3
Genomic Map