siRNA Details
| Gene Symbol: Rasgrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166493.1 | NP_001159965.1 | 6, 7 | 914 | ||
| NM_207246.4 | NP_997129.1 | 7, 8 | 906 | |||
| GenBank mRNA | AB154824.1 | - | 5, 6 | 601 | ||
| AK173041.1 | - | 6, 7 | 799 | |||
| BC066069.1 | - | 5, 6 | 544 | 6851971 | ||
| BC066127.1 | - | 7, 8 | 831 | 30280332 | ||
| CD348455.1 | - | 5, 6 | 554 | |||

Rasgrp3
Genomic Map