siRNA Details
| Gene Symbol: Pglyrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207247.4 | NP_997130.2 | 2 | 223 | ||
| GenBank mRNA | AY518698.1 | - | 2 | 194 | ||
| BB619146.1 | - | 3 | 396 | |||
| BC128291.1 | - | 2 | 194 | 40102537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pglyrp3| Gene Symbol: Pglyrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207247.4 | NP_997130.2 | 2 | 223 | ||
| GenBank mRNA | AY518698.1 | - | 2 | 194 | ||
| BB619146.1 | - | 3 | 396 | |||
| BC128291.1 | - | 2 | 194 | 40102537 | ||