siRNA Details
| Gene Symbol: E130006D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087393.1 | - | 5 | 624 | ||
| BB537478.2 | - | 3 | 376 | |||
| BC087866.1 | - | 5 | 465 | |||
| BX529972.1 | - | 4 | 353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130006D01Rik| Gene Symbol: E130006D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087393.1 | - | 5 | 624 | ||
| BB537478.2 | - | 3 | 376 | |||
| BC087866.1 | - | 5 | 465 | |||
| BX529972.1 | - | 4 | 353 | |||