siRNA Details
| Gene Symbol: Zfp532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207255.2 | NP_997138.1 | 3 | 539 | ||
| GenBank mRNA | AK083001.1 | - | 3 | 416 | ||
| BC067032.1 | - | 3 | 539 | 6841127 | ||
| BC094671.1 | - | 3 | 543 | 30618961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp532| Gene Symbol: Zfp532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207255.2 | NP_997138.1 | 3 | 539 | ||
| GenBank mRNA | AK083001.1 | - | 3 | 416 | ||
| BC067032.1 | - | 3 | 539 | 6841127 | ||
| BC094671.1 | - | 3 | 543 | 30618961 | ||