siRNA Details
| Gene Symbol: Zfp532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207255.2 | NP_997138.1 | 3 | 926 | ||
| GenBank mRNA | AK083001.1 | - | 3 | 803 | ||
| AK129410.1 | - | 4177 | ||||
| BC067032.1 | - | 3 | 926 | 6841127 | ||
| BC094671.1 | - | 3 | 930 | 30618961 | ||

Zfp532
Genomic Map