siRNA Details
| Gene Symbol: Pglyrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165968.1 | NP_001159440.1 | 7 | 936 | ||
| NM_207263.2 | NP_997146.2 | 7 | 933 | |||
| GenBank mRNA | AK137756.1 | - | 2 | 199 | ||
| AY518699.1 | - | 7 | 887 | |||
| BC120840.1 | - | 7 | 928 | 40129763 | ||
| BC144902.1 | - | 7 | 936 | 9053450 | ||
| BC144927.1 | - | 6 | 817 | |||

Pglyrp4
Genomic Map