siRNA Details
Gene Symbol: Tdpoz4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207272.2 | NP_997155.2 | 1 | 281 | ||
GenBank mRNA | BC115626.1 | - | 1 | 281 | 40102845 | |
BC115627.1 | - | 1 | 277 |
Gene Symbol: Gm4858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177694.1 | NP_001171165.1 | 1 | 281 | ||
GenBank mRNA | BC172161.1 | - | 1 | 265 |
