siRNA Details
| Gene Symbol: Tdpoz4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207272.2 | NP_997155.2 | 1 | 334 | ||
| GenBank mRNA | BC115626.1 | - | 1 | 334 | 40102845 | |
| BC115627.1 | - | 1 | 330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdpoz4| Gene Symbol: Tdpoz4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207272.2 | NP_997155.2 | 1 | 334 | ||
| GenBank mRNA | BC115626.1 | - | 1 | 334 | 40102845 | |
| BC115627.1 | - | 1 | 330 | |||