siRNA Details
| Gene Symbol: Tdpoz5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207273.2 | NP_997156.2 | 211 | |||
| GenBank mRNA | BC111104.1 | - | 1 | 209 | ||
| Gene Symbol: Gm10697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163731.1 | NP_001157203.1 | 1 | 211 | ||

Tdpoz5
Genomic Map