siRNA Details
| Gene Symbol: REEP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005669.4 | NP_005660.4 | 5 | 1031 | ||
| GenBank mRNA | AK295436.1 | - | 4 | 783 | ||
| AK298268.1 | - | 4 | 766 | |||
| AK300048.1 | - | 5 | 849 | |||
| BC000232.1 | - | 5 | 912 | 3350749 | ||
| BX648005.1 | - | 5 | 1012 | |||
| M74090.1 | - | 5 | 913 | |||

REEP5
Genomic Map