siRNA Details
| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207301.2 | NP_997184.1 | 5 | 1347 | ||
| GenBank mRNA | AK172079.1 | - | 5 | 1353 | ||
| BC031769.1 | - | 5 | 1340 | 4235618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wrb| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207301.2 | NP_997184.1 | 5 | 1347 | ||
| GenBank mRNA | AK172079.1 | - | 5 | 1353 | ||
| BC031769.1 | - | 5 | 1340 | 4235618 | ||