siRNA Details
| Gene Symbol: BC049702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049702.1 | - | 2 | 804 | 6707281 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC049702| Gene Symbol: BC049702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049702.1 | - | 2 | 804 | 6707281 | |