siRNA Details
Gene Symbol: 4930504O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207527.3 | NP_997410.1 | 4 | 410 | ||
GenBank mRNA | AK076838.1 | - | 4 | 368 | ||
BC048595.1 | - | 4 | 371 | 6740346 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930504O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207527.3 | NP_997410.1 | 4 | 410 | ||
GenBank mRNA | AK076838.1 | - | 4 | 368 | ||
BC048595.1 | - | 4 | 371 | 6740346 |