siRNA Details
| Gene Symbol: Brinp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207583.2 | NP_997466.2 | 6 | 1661 | ||
| GenBank mRNA | AK032297.1 | - | 6 | 1665 | ||
| AK045801.1 | - | 6 | 1188 | |||
| AK122549.1 | - | 6 | 1727 | |||
| BC067038.1 | - | 6 | 2466 | |||
| BC076572.1 | - | 6 | 1661 | 30363878 | ||

Brinp2
Genomic Map