siRNA Details
| Gene Symbol: Olfr553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207621.1 | NP_997504.1 | 1 | 428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr553| Gene Symbol: Olfr553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207621.1 | NP_997504.1 | 1 | 428 | ||