siRNA Details
Gene Symbol: Hook3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207659.3 | NP_997542.1 | 9 | 894 | ||
GenBank mRNA | AK054263.1 | - | 9 | 813 | ||
AK080377.1 | - | 9 | 861 | |||
AK145149.1 | - | 9 | 864 | |||
AY223806.1 | - | 9 | 684 | |||
BC090960.1 | - | 9 | 804 | 30651062 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
