siRNA Details
| Gene Symbol: Hoxc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010464.2 | NP_034594.1 | 2 | 1358 | ||
| GenBank mRNA | AF193796.1 | - | 2 | 1358 | ||
| AY349616.1 | - | 2 | 1386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc13| Gene Symbol: Hoxc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010464.2 | NP_034594.1 | 2 | 1358 | ||
| GenBank mRNA | AF193796.1 | - | 2 | 1358 | ||
| AY349616.1 | - | 2 | 1386 | |||