siRNA Details
| Gene Symbol: SNUPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042581.1 | NP_001036046.1 | 5 | 800 | ||
| NM_001042588.1 | NP_001036053.1 | 5 | 605 | |||
| NM_005701.3 | NP_005692.1 | 5 | 905 | |||
| GenBank mRNA | AF039029.1 | - | 5 | 550 | ||
| AK054641.1 | - | 6 | 1038 | |||
| AK289475.1 | - | 5 | 601 | |||
| BC004203.2 | - | 5 | 536 | 3605328 | ||
| CR456811.1 | - | 4 | 460 | |||

SNUPN
Genomic Map