siRNA Details
| Gene Symbol: Myo18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291212.1 | NP_001278141.1 | 8 | 1893 | ||
| NM_001291213.1 | NP_001278142.1 | 8 | 2001 | |||
| NM_001291214.1 | NP_001278143.1 | 7 | 860 | |||
| NM_001291215.1 | NP_001278144.1 | 8 | 896 | |||
| NM_011586.3 | NP_035716.1 | 7 | 1857 | |||
| GenBank mRNA | AB026497.1 | - | 7 | 1827 | ||
| AK137574.1 | - | 7 | 848 | |||
| AK147584.1 | - | 8 | 845 | |||
| AK171342.1 | - | 7 | 857 | |||
| BC138366.1 | - | 8 | 1835 | 8861386 | ||
| BC145257.1 | - | 8 | 1943 | |||

Myo18a
Genomic Map