siRNA Details
| Gene Symbol: Myo18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291212.1 | NP_001278141.1 | 41 | 7337 | ||
| NM_001291213.1 | NP_001278142.1 | 41 | 7445 | |||
| NM_001291214.1 | NP_001278143.1 | 39 | 6292 | |||
| NM_001291215.1 | NP_001278144.1 | 42 | 6358 | |||
| NM_011586.3 | NP_035716.1 | 40 | 7301 | |||
| GenBank mRNA | AB026497.1 | - | 40 | 7270 | ||
| AK147584.1 | - | 42 | 6307 | |||
| BC046638.1 | - | 23 | 4129 | |||
| BC057920.1 | - | 11 | 2512 | |||

Myo18a
Genomic Map