siRNA Details
| Gene Symbol: Reg3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161741.1 | NP_001155213.1 | 5 | 449 | ||
| NM_013893.2 | NP_038921.2 | 6 | 587 | |||
| GenBank mRNA | AB028625.1 | - | 6 | 593 | ||
| AB035204.1 | - | 6 | 567 | |||
| AK019033.1 | - | 6 | 566 | |||
| BC113760.1 | - | 6 | 582 | 40055351 | ||
| BM055959.1 | - | 4 | 397 | |||

Reg3d
Genomic Map