siRNA Details
| Gene Symbol: Siva1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161737.1 | NP_001155209.1 | 3 | 495 | ||
| NM_013929.2 | NP_038957.2 | 4 | 690 | |||
| GenBank mRNA | AF033114.1 | - | 4 | 638 | ||
| AK010562.1 | - | 4 | 616 | |||
| AK017570.1 | - | 4 | 651 | |||
| AK153115.1 | - | 4 | 600 | |||
| AK169200.1 | - | 4 | 643 | |||
| AK194303.1 | - | 2 | 117 | |||
| AK207814.1 | - | 2 | 117 | |||
| AK218153.1 | - | 2 | 117 | |||
| BC012279.1 | - | 4 | 672 | 4013026 | ||

Siva1
Genomic Map