siRNA Details
| Gene Symbol: Fam60a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019643.3 | NP_062617.2 | 6 | 1367 | ||
| GenBank mRNA | AK046519.1 | - | 6 | 1276 | ||
| AK145384.1 | - | 6 | 1378 | |||
| AK166569.1 | - | 6 | 1373 | |||
| BC076599.1 | - | 6 | 1367 | 30614283 | ||
| U64033.1 | - | 6 | 1459 | |||

Fam60a
Genomic Map