siRNA Details
| Gene Symbol: Jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021310.3 | NP_067285.2 | 2 | 1527 | ||
| GenBank mRNA | AB221690.1 | - | 2 | 1025 | ||
| AF201390.1 | - | 2 | 1151 | |||
| AK139934.1 | - | 2 | 1156 | |||
| AK141957.1 | - | 2 | 1528 | |||
| BC020052.1 | - | 2 | 1452 | |||
| BC069906.1 | - | 2 | 1381 | |||
| BC090835.1 | - | 2 | 1408 | 5705427 | ||

Jmy
Genomic Map