siRNA Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025590.4 | NP_079866.2 | 3, 4 | 429 | ||
| GenBank mRNA | AF416923.1 | - | 3, 4 | 367 | ||
| AK031588.1 | - | 3, 4 | 346 | |||
| AK038546.1 | - | 3, 4 | 367 | |||
| AK048670.1 | - | 3, 4 | 429 | |||
| AK052332.1 | - | 3, 4 | 445 | |||
| AK122350.1 | - | 2, 3 | 216 | |||
| BC042492.1 | - | 3, 4 | 364 | 4240966 | ||

Acot11
Genomic Map