siRNA Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025590.4 | NP_079866.2 | 16 | 5543 | ||
| GenBank mRNA | AK003873.1 | - | 1 | 941 | ||
| AK122350.1 | - | 15 | 5290 | |||
| BC042492.1 | - | 16 | 5483 | 4240966 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acot11| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025590.4 | NP_079866.2 | 16 | 5543 | ||
| GenBank mRNA | AK003873.1 | - | 1 | 941 | ||
| AK122350.1 | - | 15 | 5290 | |||
| BC042492.1 | - | 16 | 5483 | 4240966 | ||