siRNA Details
| Gene Symbol: Fam166a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026624.3 | NP_080900.1 | 3 | 344 | ||
| GenBank mRNA | AK016459.1 | - | 3 | 346 | ||
| BC061039.1 | - | 3 | 347 | 6704633 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam166a| Gene Symbol: Fam166a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026624.3 | NP_080900.1 | 3 | 344 | ||
| GenBank mRNA | AK016459.1 | - | 3 | 346 | ||
| BC061039.1 | - | 3 | 347 | 6704633 | ||