siRNA Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029012.2 | NP_083288.2 | 11 | 1614 | ||
| GenBank mRNA | AK014709.1 | - | 11 | 1952 | ||
| BC023131.1 | - | 7 | 1229 | |||
| BC078437.1 | - | 2 | 1661 | |||
| BC086646.1 | - | 11 | 1979 | 6817386 | ||

Sppl3
Genomic Map